Anti-N6AMT1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: N6AMT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-626
- 77856-819
- Antigen symbol:
- N6AMT1
- N6AMT1
- Antigen synonyms:
- N6AMT1|C21orf127|HEMK2|PRED28|Methyltransferase N6AMT1|HemK methyltransferase family member 2|M.HsaHemK2P|Methylarsonite methyltransferase N6AMT1|EC 2.1.1.-|N(6)-adenine-specific DNA methyltransferase 1|EC 2.1.1.72|Protein N(5)-glutamine methyltransferase
- N6AMT1|C21orf127|HEMK2|PRED28|Methyltransferase N6AMT1|HemK methyltransferase family member 2|M.HsaHemK2P|Methylarsonite methyltransferase N6AMT1|EC 2.1.1.-|N(6)-adenine-specific DNA methyltransferase 1|EC 2.1.1.72|Protein N(5)-glutamine methyltransferase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5N5
- Q9Y5N5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human HemK methyltransferase family member 2 protein (1-186AA)
- Recombinant Human HemK methyltransferase family member 2 protein (1-186AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together