Anti-GNAT1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GNAT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-552
- 77856-185
- Antigen symbol:
- GNAT1
- GNAT1
- Antigen synonyms:
- CSNBAD3|rod-specific|guanine nucleotide binding protein G(T) alpha 1 subunit|Guanine nucleotide-binding protein G(t) subunit alpha-1|Transducin alpha-1 chain|GNATR|rod-type transducin alpha subunit|GNAT1_HUMAN|GBT1|Rod specific transducin|transducin|guanine nucleotide binding protein (G protein) alpha transducing activity polypeptide 1
- CSNBAD3|GBT1|GNAT1_HUMAN|GNATR|guanine nucleotide binding protein (G protein) alpha transducing activity polypeptide 1|guanine nucleotide binding protein G(T) alpha 1 subunit|Guanine nucleotide-binding protein G(t) subunit alpha-1|Rod specific transducin|rod-type transducin alpha subunit|Transducin alpha-1 chain|transducin|rod-specific
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P11488
- P11488
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Guanine nucleotide-binding protein G(t) subunit alpha-1 protein (92-170AA)
- Recombinant Human Guanine nucleotide-binding protein G(t) subunit alpha-1 protein (92-170AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together