Anti-PSMB4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PSMB4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-056
- 77859-699
- Antigen symbol:
- PSMB4
- PSMB4
- Antigen synonyms:
- 26 kDa prosomal protein|HN3|HsBPROS26|HSN3|Macropain beta chain|Multicatalytic endopeptidase complex beta chain|PROS-26|PROS26|Proteasome (prosome macropain) subunit beta type 4|Proteasome beta 4 subunit|Proteasome beta chain|Proteasome chain 3|Proteasome subunit beta 4|Proteasome subunit beta type 4|Proteasome subunit beta type-4|Proteasome subunit HsN3|PSB4_HUMAN|PSMB 4|PSMB4
- 26 kDa prosomal protein|HN3|HsBPROS26|HSN3|Macropain beta chain|Multicatalytic endopeptidase complex beta chain|PROS-26|PROS26|Proteasome (prosome macropain) subunit beta type 4|Proteasome beta 4 subunit|Proteasome beta chain|Proteasome chain 3|Proteasome subunit beta 4|Proteasome subunit beta type 4|Proteasome subunit beta type-4|Proteasome subunit HsN3|PSB4_HUMAN|PSMB 4|PSMB4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P28070
- P28070
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Proteasome subunit beta type-4 protein (46-264AA)
- Recombinant Human Proteasome subunit beta type-4 protein (46-264AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together