Anti-LGR6 Rabbit Polyclonal Antibody
Catalogue # 77742-131
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:LGR6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9HBX8
- Antigen synonyms:FLJ14471|Gonadotropin receptor|GPCR|Leucine rich repeat containing G protein coupled receptor 6|Leucine-rich repeat-containing G-protein coupled receptor 6|LGR6_HUMAN|VTS20631
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:VTS20631
- Purification:Antigen affinity purified
- Packaging:Vial
Spécifications
À propos de cet article
Type:
Antigen: LGR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat