Anti-LCP1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LCP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-865
- 77742-028
- Antigen symbol:
- LCP1
- LCP1
- Antigen synonyms:
- CP64| L plastin| L-plastin| Larval cuticle protein 1| LC64P| LCP-1| LCP1| LPL| Lplastin| Lymphocyte cytosolic protein 1| Plastin 2| Plastin-2| PLS2| PLSL_HUMAN
- CP64|L plastin|L-plastin|Larval cuticle protein 1|LC64P|LCP-1|LCP1|LPL|Lplastin|Lymphocyte cytosolic protein 1|Plastin 2|Plastin-2|PLS2|PLSL_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P13796
- P13796
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Plastin-2 protein (2-627AA)
- Recombinant Human Plastin-2 protein (2-627AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together