Anti-HJURP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HJURP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-088
- 77857-446
- Antigen symbol:
- HJURP
- HJURP
- Antigen synonyms:
- FAKTS|FLEG1|URLC9|Holliday junction recognition protein|14-3-3-associated AKT substrate|Fetal liver-expressing gene 1 protein|Up-regulated in lung cancer 9
- FAKTS|FLEG1|URLC9|Holliday junction recognition protein|14-3-3-associated AKT substrate|Fetal liver-expressing gene 1 protein|Up-regulated in lung cancer 9
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NCD3
- Q8NCD3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Holliday junction recognition protein (1-748AA)
- Recombinant human Holliday junction recognition protein (1-748AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together