Anti-DEFA3 Rabbit Polyclonal Antibody
Catalog # 77736-029
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:DEFA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P59666
- Antigen synonyms:DEF3Neutrophil defensin 3|Defensin|alpha 3|HNP-3|HP-3|HP3) [Cleaved into: HP 3-56|Neutrophil defensin 2|HNP-2|HP-2|HP2)]
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:HP2
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:20000
Type:
Antigen: DEFA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human