Anti-LENG1 Rabbit Polyclonal Antibody
Catalog # 77742-076
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:LENG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96BZ8
- Antigen synonyms:Leukocyte receptor cluster member 1
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Leukocyte receptor cluster member 1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: LENG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat