Order Entry
Canada
ContactUsLinkComponent
Anti-ETS2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-ETS2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-ETS2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77854-969
  • 77736-237
  • Antigen symbol:
  • ETS2
  • ETS2
  • Antigen synonyms:
  • C ets 2 protein|ETS2_HUMAN|ETS2IT1|Oncogene ETS 2|Oncogene ETS2|Protein C-ets-2|v ets avian erythroblastosis virus E2 oncogene homolog 2|V ets avian erythroblastosis virus E26 oncogene homolog 2|V ets erythroblastosis virus E26 oncogene homolog 2
  • C ets 2 protein|ETS2_HUMAN|ETS2IT1|Oncogene ETS 2|Oncogene ETS2|Protein C-ets-2|v ets avian erythroblastosis virus E2 oncogene homolog 2|V ets avian erythroblastosis virus E26 oncogene homolog 2|V ets erythroblastosis virus E26 oncogene homolog 2
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P15036
  • P15036
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Protein C-ets-2 protein (9-185AA)
  • Recombinant Human Protein C-ets-2 protein (9-185AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together