Anti-IFIT1 Rabbit Recombinant Polyclonal Antibody
77740-632
New Product
- Antigen symbol:IFIT1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P09914
- Antigen synonyms:C56 antibody|G10P1 antibody|GARG 16 antibody|GARG16 antibody|IFI 56 antibody|IFI 56K antibody|IFI-56K antibody|IFI56 antibody|IFIT-1 antibody|Ifit1 antibody|IFIT1_HUMAN antibody|IFNAI1 antibody|Interferon alpha inducible protein (MW 56kD) antibody|Interferon induced 56 kDa protein antibody|Interferon induced protein 56 antibody|Interferon inducible mRNA 561 antibody|Interferon-induced 56 kDa protein antibody|Interferon-induced protein with tetratricopeptide repeats 1 antibody|ISG56 antibody|OTTHUMP00000020062 antibody|P56 antibody|RNM561 antibody
- Storage buffer:PBS with 0.1% sodium azide, 50% glycerol, pH 7.3. –20 °C, Avoid freeze / thaw cycles.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Interferon-induced protein with tetratricopeptide repeats 1 protein (1-181AA)
- Purification:Antigen Affinity Purified
- Packaging:Vial
Type:
Antigen: IFIT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat