Anti-FCN1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: FCN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-274
- 77737-003
- Antigen symbol:
- FCN1
- FCN1
- Antigen synonyms:
- Collagen/fibrinogen domain containing protein 1|Collagen/fibrinogen domain-containing protein 1|Fcn1|FCN1_HUMAN|FCNM|ficolin (collagen/fibrinogen domain containing) 1|Ficolin 1|Ficolin A|Ficolin alpha|Ficolin-1|Ficolin-A|Ficolin-alpha|M ficolin|M-ficolin
- Collagen/fibrinogen domain containing protein 1|Collagen/fibrinogen domain-containing protein 1|Fcn1|FCN1_HUMAN|FCNM|ficolin (collagen/fibrinogen domain containing) 1|Ficolin 1|Ficolin A|Ficolin alpha|Ficolin-1|Ficolin-A|Ficolin-alpha|M ficolin|M-ficolin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00602
- O00602
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ficolin-1 protein (30-326AA)
- Recombinant Human Ficolin-1 protein (30-326AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together