Anti-FZD9 Rabbit Polyclonal Antibody
Catalog # 77737-781
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:FZD9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O00144
- Antigen synonyms:Frizzled Drosophila homolog of 3|Fz-9|FZD3|Frizzled homolog 9|FzE6|Xfz9|FZD9_HUMAN|FZD 3|Frizzled-9|mfz9|Fz 9|hFz9|Frizzled homolog 9 (Drosophila)|CD 349 antigen|CD349|Fzd 9|CD 349|FZD3-PEN
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Xfz9
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: FZD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat