Anti-MYH3 Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MYH3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-732
- 77860-196
- Antigen symbol:
- MYH3
- MYH3
- Antigen synonyms:
- embryonic|fast skeletal muscle|HEMHC|Muscle embryonic myosin heavy chain 3|Muscle embryonic myosin heavy chain|MYH 3|MYH3_HUMAN|MYHC EMB|MYHSE 1|MYHSE1|Myosin heavy chain 3|Myosin heavy chain 3 skeletal muscle embryonic|Myosin heavy chain|Myosin heavy chain fast skeletal muscle embryonic|Myosin Heavy Polypeptide 3|Myosin heavy polypeptide 3 skeletal muscle embryonic|Myosin skeletal heavy chain embryonic 1|Myosin-3|SMHCE
- embryonic|fast skeletal muscle|HEMHC|Muscle embryonic myosin heavy chain 3|Muscle embryonic myosin heavy chain|MYH 3|MYH3_HUMAN|MYHC EMB|MYHSE 1|MYHSE1|Myosin heavy chain 3|Myosin heavy chain 3 skeletal muscle embryonic|Myosin heavy chain|Myosin heavy chain fast skeletal muscle embryonic|Myosin Heavy Polypeptide 3|Myosin heavy polypeptide 3 skeletal muscle embryonic|Myosin skeletal heavy chain embryonic 1|Myosin-3|SMHCE
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P11055
- P11055
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Myosin-3 protein (1-217AA)
- Recombinant human Myosin-3 protein (1-217AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together