Anti-FOXD4 Rabbit Polyclonal Antibody
77736-733
New Product
- Antigen symbol:FOXD4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q12950
- Antigen synonyms:FKHL9|Forkhead box D4|Forkhead box protein D4|Forkhead Drosophila homolog like 9|Forkhead like 9|Forkhead related activator 5|Forkhead related protein FKHL9|Forkhead related transcription factor 5|Forkhead-related protein FKHL9|Forkhead-related transcription factor 5|FOXD4|FOXD4_HUMAN|FOXD4A|FREAC 5|FREAC-5|FREAC5|MGC105106|Myeloid factor alpha|Myeloid factor-alpha
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:FKHL9
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB:1:500 to 1:2000, ELISA:1:10000
Type:
Antigen: FOXD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse