Anti-KLF11 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KLF11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-726
- 77858-630
- Antigen symbol:
- KLF11
- KLF11
- Antigen synonyms:
- TIEG-2|D12Ertd427e|TIEG-3|Transforming Growth Factor Beta Inducible Early Growth Response 2|MODY7|Transforming growth factor-beta-inducible early growth response protein 3|Krueppel like factor 11|Transforming growth factor-beta-inducible early growth response protein 2|TGFB Early Growth Response 2|TGFB-inducible early growth response protein 3|Tieg2b|FKLF1|TGFB-inducible early growth response protein 2|Krueppel-like transcription factor 1|TGFB-inducible early growth response protein 2b|Tcfcp2l2|TIEG 2|KLF11_HUMAN|Krueppel-like factor 11|9830142A17|FKLF
- 9830142A17|D12Ertd427e|FKLF|FKLF1|KLF11_HUMAN|Krueppel like factor 11|Krueppel-like factor 11|Krueppel-like transcription factor 1|MODY7|Tcfcp2l2|TGFB Early Growth Response 2|TGFB-inducible early growth response protein 2|TGFB-inducible early growth response protein 2b|TGFB-inducible early growth response protein 3|TIEG 2|TIEG-2|TIEG-3|Tieg2b|Transforming Growth Factor Beta Inducible Early Growth Response 2|Transforming growth factor-beta-inducible early growth response protein 2|Transforming growth factor-beta-inducible early growth response protein 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14901
- O14901
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Krueppel-like factor 11 protein (107-205AA)
- Recombinant Human Krueppel-like factor 11 protein (107-205AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together