Anti-LETM1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LETM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-910
- 77742-089
- Antigen symbol:
- LETM1
- LETM1
- Antigen synonyms:
- LETM 1| LETM1 and EF hand domain containing protein 1| mitochondrial| LETM1 and EF-hand domain-containing protein 1| Letm1| LETM1_HUMAN| Leucine zipper EF hand containing transmembrane protein 1| Leucine zipper-EF-hand-containing transmembrane protein 1| Mdm38 homolog
- LETM 1|LETM1 and EF hand domain containing protein 1|mitochondrial|LETM1 and EF-hand domain-containing protein 1|Letm1|LETM1_HUMAN|Leucine zipper EF hand containing transmembrane protein 1|Leucine zipper-EF-hand-containing transmembrane protein 1|Mdm38 homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95202
- O95202
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mitochondrial proton/calcium exchanger protein (443-543AA)
- Recombinant Human Mitochondrial proton/calcium exchanger protein (443-543AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together