Anti-GPR12 Rabbit Polyclonal Antibody
Catalog # 77737-544
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:GPR12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P47775
- Antigen synonyms:FLJ18149|FLJ97704|G protein coupled receptor 12|G-protein coupled receptor 12|GPCR12|GPCR21|GPR 12|Gpr12|GPR12_HUMAN|MGC138349|MGC138351|Probable G protein coupled receptor GPR12
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:FLJ18149
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IF: 1:200 to 1:1000, ELISA: 1:5000
Type:
Antigen: GPR12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human