Anti-INMT Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: INMT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-088
- 77858-115
- Antigen symbol:
- INMT
- INMT
- Antigen synonyms:
- Amine N methyltransferase|Amine N-methyltransferase|Aromatic alkylamine N methyltransferase|Aromatic alkylamine N-methyltransferase|Arylamine N methyltransferase|Arylamine N-methyltransferase|Indolamine N-methyltransferase|Indolethylamine N methyltransferase|Indolethylamine N-methyltransferase|Inmt|INMT_HUMAN|nicotine N methyltransferase |TEMT|thioester S methyltransferase like|Thioether S methyltransferase
- Amine N methyltransferase| Amine N-methyltransferase| Aromatic alkylamine N methyltransferase| Aromatic alkylamine N-methyltransferase| Arylamine N methyltransferase| Arylamine N-methyltransferase| Indolamine N-methyltransferase| Indolethylamine N methyltransferase| Indolethylamine N-methyltransferase| Inmt| INMT_HUMAN| nicotine N methyltransferase | TEMT| thioester S methyltransferase like| Thioether S methyltransferase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95050
- O95050
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Indolethylamine N-methyltransferase protein (1-263AA)
- Recombinant Human Indolethylamine N-methyltransferase protein (1-263AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together