Anti-MVD Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MVD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77860-134
- 77743-655
- Antigen symbol:
- MVD
- MVD
- Antigen synonyms:
- Diphosphomevalonate decarboxylase|FP17780|MDDase|Mevalonate (diphospho) decarboxylase|Mevalonate (diphospho)decarboxylase|Mevalonate pyrophosphate decarboxylase|MPD|Mvd|MVD1_HUMAN
- Diphosphomevalonate decarboxylase|FP17780|MDDase|Mevalonate (diphospho) decarboxylase|Mevalonate (diphospho)decarboxylase|Mevalonate pyrophosphate decarboxylase|MPD|Mvd|MVD1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P53602
- P53602
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Diphosphomevalonate decarboxylase protein (69-221AA)
- Recombinant Human Diphosphomevalonate decarboxylase protein (69-221AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together