Order Entry
Canada
ContactUsLinkComponent
Anti-KDM4A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-KDM4A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-KDM4A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77858-475
  • 77741-541
  • Antigen symbol:
  • KDM4A
  • KDM4A
  • Antigen synonyms:
  • JHDM3A| JmjC domain containing histone demethylation protein 3A| JmjC domain-containing histone demethylation protein 3A| JMJD2| JMJD2A| jumonji C domain containing histone demethylase 3A | Jumonji domain containing 2| Jumonji domain containing 2A| Jumonji domain containing protein 2A| Jumonji domain-containing protein 2A| KDM4A| KDM4A_HUMAN| KIAA0677| Lysine (K) specific demethylase 4A| Lysine-specific demethylase 4A| TDRD14A| Tudor domain containing 14A
  • JHDM3A|JmjC domain containing histone demethylation protein 3A|JmjC domain-containing histone demethylation protein 3A|JMJD2|JMJD2A|jumonji C domain containing histone demethylase 3A |Jumonji domain containing 2|Jumonji domain containing 2A|Jumonji domain containing protein 2A|Jumonji domain-containing protein 2A|KDM4A|KDM4A_HUMAN|KIAA0677|Lysine (K) specific demethylase 4A|Lysine-specific demethylase 4A|TDRD14A|Tudor domain containing 14A
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O75164
  • O75164
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Lysine-specific demethylase 4A protein (127-364AA)
  • Recombinant Human Lysine-specific demethylase 4A protein (127-364AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together