Anti-MTTP Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: MTTP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77860-117
- 77743-629
- Antigen symbol:
- MTTP
- MTTP
- Antigen synonyms:
- ABL|Microsomal triglyceride transfer protein (large polypeptide 88kDa)|Microsomal triglyceride transfer protein 88kD|Microsomal triglyceride transfer protein|Microsomal triglyceride transfer protein large subunit|MTP|MTP large subunit|MTP_HUMAN|Transfer RNA|mitochondrial|proline|tRNA-Pro
- ABL|Microsomal triglyceride transfer protein (large polypeptide 88kDa)|Microsomal triglyceride transfer protein 88kD|Microsomal triglyceride transfer protein|Microsomal triglyceride transfer protein large subunit|MTP|MTP large subunit|MTP_HUMAN|Transfer RNA|mitochondrial|proline|tRNA-Pro
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55157
- P55157
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Microsomal triglyceride transfer protein large subunit protein (19-151AA)
- Recombinant Human Microsomal triglyceride transfer protein large subunit protein (19-151AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together