Anti-HMGCL Rabbit Polyclonal Antibody
Catalog # 77740-175
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:HMGCL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P35914
- Antigen synonyms:3 hydroxy 3 methylglutaryl CoA lyase|3 hydroxy 3 methylglutaryl Coenzyme A lyase|3 hydroxymethyl 3 methylglutaryl Coenzyme A lyase (hydroxymethylglutaricaciduria)|3 hydroxymethyl 3 methylglutaryl Coenzyme A lyase|3-hydroxy-3-methylglutarate-CoA lyase|HL|HMG CoA lyase|HMG CoA Lyase Deficiency|HMG-CoA lyase|HMGCL|HMGCL_HUMAN|Hydroxymethylglutaricaciduria|Hydroxymethylglutaryl CoA lyase|Hydroxymethylglutaryl CoA lyase mitochondrial|Hydroxymethylglutaryl-CoA lyase|Mitochondrial 3 hydroxy 3 methylglutaryl CoA lyase|mitochondrial|MS725|OTTHUMP00000044830
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:OTTHUMP00000044830
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: HMGCL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat