Anti-NAT2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NAT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-955
- 77743-954
- Antigen symbol:
- NAT2
- NAT2
- Antigen synonyms:
- AAC2|ARY2_HUMAN|Arylamide acetylase 2 (N-acetyltransferase 2, isoniazid inactivation)|Arylamide acetylase 2|Arylamine N-acetyltransferase 2|HGNC:7646|N-acetyltransferase 2 (arylamine N-acetyltransferase)|N-acetyltransferase type 2|NAT-2|NAT2|PNAT|Polymorphic arylamine N-acetyltransferase
- AAC2|ARY2_HUMAN|Arylamide acetylase 2 (N-acetyltransferase 2, isoniazid inactivation)|Arylamide acetylase 2|Arylamine N-acetyltransferase 2|HGNC:7646|N-acetyltransferase 2 (arylamine N-acetyltransferase)|N-acetyltransferase type 2|NAT-2|NAT2|PNAT|Polymorphic arylamine N-acetyltransferase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P11245
- P11245
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Arylamine N-acetyltransferase 2 protein (1-290AA)
- Arylamine N-acetyltransferase 2 protein (1-290AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together