Anti-FGFRL1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: FGFRL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77855-400
- 77737-175
- Antigen symbol:
- FGFRL1
- FGFRL1
- Antigen synonyms:
- FGF receptor like protein 1|Fibroblast growth factor receptor-like 1|Fgfrl1|FGF homologous factor receptor|FGFR 5|FGRL1_HUMAN|FHFR|FGFR-5|FGFR like protein|Fibroblast growth factor receptor 5|FGF receptor-like protein 1|FGFR-like protein|FGFR5|Fibroblast growth factor receptor like 1
- FGF homologous factor receptor|FGF receptor like protein 1|FGF receptor-like protein 1|FGFR 5|FGFR like protein|FGFR-5|FGFR-like protein|FGFR5|Fgfrl1|FGRL1_HUMAN|FHFR|Fibroblast growth factor receptor 5|Fibroblast growth factor receptor like 1|Fibroblast growth factor receptor-like 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N441
- Q8N441
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Fibroblast growth factor receptor-like 1 protein (403-470AA)
- Recombinant Human Fibroblast growth factor receptor-like 1 protein (403-470AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together