Order Entry
Canada
ContactUsLinkComponent
12303
12303
Catalog # 77737-595
Supplier:  AFG Bioscience
New Product
12303
Catalog # 77737-595
Supplier:  AFG Bioscience
Supplier Number:  A22406-50UG
New Product

Specifications

  • Size:
    50 µg

Specifications

About this item

Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:20000

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human