Anti-E2F5 Rabbit Polyclonal Antibody
Catalog # 77736-268
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:E2F5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q15329
- Antigen synonyms:E2F 5|E2F transcription factor 5|E2F transcription factor 5 p130 binding|E2F-5|E2f5|E2F5_HUMAN|p130 binding|Transcription factor E2F5
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:E2F 5
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:2000, ELISA:1:20000
Type:
Antigen: E2F5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat