Anti-LCT Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LCT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-036
- 77858-874
- Antigen symbol:
- LCT
- LCT
- Antigen synonyms:
- LAC|Lactase|Lactase phlorizin hydrolase 1|Lactase phlorizin hydrolase|Lactase-glycosylceramidase|Lct|LPH|LPH_HUMAN|LPH1|Phlorizin hydrolase
- LAC|Lactase|Lactase phlorizin hydrolase 1|Lactase phlorizin hydrolase|Lactase-glycosylceramidase|Lct|LPH|LPH_HUMAN|LPH1|Phlorizin hydrolase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P09848
- P09848
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lactase-phlorizin hydrolase protein (983-1252AA)
- Recombinant Human Lactase-phlorizin hydrolase protein (983-1252AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together