Anti-IL11 Rabbit Polyclonal Antibody
Catalogue # 77738-501
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:IL11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:P20809
- Antigen synonyms:Adipogenesis inhibitory factor| AGIF| IL 11| IL-11| IL11_HUMAN| Interleukin 11| Interleukin-11| Oprelvekin
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:Shipped on Blue ice
- Immunogen:Adipogenesis inhibitory factor
- Purification:Antigen affinity purified
- Packaging:Vial
Spécifications
À propos de cet article
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: IL11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat