Anti-S1PR1 Rabbit Polyclonal Antibody
Catalog # 77736-279
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:S1PR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P21453
- Antigen synonyms:CD363|CHEDG 1|CHEDG1|D1S3362|ECGF 1|ECGF1|EDG 1|EDG1|endothelial differentiation G protein coupled receptor 1|Endothelial differentiation G-protein coupled receptor 1|Endothelial differentiation sphingolipid G protein coupled receptor 1|FLJ58121|G protein coupled sphingolipid receptor|g protein-coupled receptor edg-1|S1P receptor 1|S1P receptor Edg 1|S1P receptor Edg-1|S1P receptor Edg1|S1P(1) receptor|S1P1|S1PR1_HUMAN|sphingolipid g-protein-coupled receptor 1|Sphingosine 1 phosphate receptor Edg 1|Sphingosine 1 phosphate receptor EDG1|sphingosine 1- phosphate receptor 1|Sphingosine 1-phosphate receptor 1|Sphingosine 1-phosphate receptor Edg-1|Sphingosine 1phosphate receptor type 1 S1P1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:CD363
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IF: 1:200 to 1:1000, ELISA: 1:10000
Type:
Antigen: S1PR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat