Anti-NFIC Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NFIC
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-389
- 77744-388
- Antigen symbol:
- NFIC
- NFIC
- Antigen synonyms:
- CCAAT box binding transcription factor|Nuclear factor 1/C|Nuclear factor 1 C type|TGGCA-binding protein|NF1-C|1500041O16Rik|AA589446|AI746521|NF-I/C|CTF|MGC137374|CCAAT binding transcription factor|NFIC_HUMAN|NFI-C|Nuclear factor 1|CTF5|NFI/C|CAAT box transcription factor|MGC20153|NF I|Nuclear factor 1 C-type|TGGCA binding protein|Nuclear factor I/C (CCAAT binding transcription factor)|NF1C|Nuclear factor I/C|CCAAT-box-binding transcription factor|Transcription factor NFIC|CNFI C|NF1 C|1110019L22Rik|NF I/C|NFI
- CCAAT box binding transcription factor|Nuclear factor 1/C|Nuclear factor 1 C type|TGGCA-binding protein|NF1-C|1500041O16Rik|AA589446|AI746521|NF-I/C|CTF|MGC137374|CCAAT binding transcription factor|NFIC_HUMAN|NFI-C|Nuclear factor 1|CTF5|NFI/C|CAAT box transcription factor|MGC20153|NF I|Nuclear factor 1 C-type|TGGCA binding protein|Nuclear factor I/C (CCAAT binding transcription factor)|NF1C|Nuclear factor I/C|CCAAT-box-binding transcription factor|Transcription factor NFIC|CNFI C|NF1 C|1110019L22Rik|NF I/C|NFI
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P08651
- P08651
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nuclear factor 1 C-type protein (201-428AA)
- Nuclear factor 1 C-type protein (201-428AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together