Anti-HSD17B6 Rabbit Polyclonal Antibody
Catalog # 77740-427
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:HSD17B6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O14756
- Antigen synonyms:17-beta-HSD 6|17-beta-HSD6|17-beta-hydroxysteroid dehydrogenase type 6|3 hydroxysteroid epimerase|3(alpha >beta) hydroxysteroid epimerase|3(alpha >beta) hydroxysteroid epimerasel|3-alpha->,beta-HSE|3-alpha->,beta-hydroxysteroid epimerase|H17B6_HUMAN|HSE|Hydroxysteroid (17 beta) dehydrogenase 6|Hydroxysteroid (17 beta) dehydrogenase 6 homolog (mouse)|Hydroxysteroid 17 beta dehydrogenase 6|NAD+ dependent 3 alpha hydroxysteroid dehydrogenase 3 hydroxysteroid epimerase|NAD+ dependent 3 alpha hydroxysteroid dehydrogenase|Oxidative 3 alpha hydroxysteroid dehydrogenase|Oxidative 3-alpha hydroxysteroid dehydrogenase|Oxidoreductase|Retinol dehydrogenase|RODH|SDR9C6|Short chain dehydrogenase/reductase family 9C|member 6
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Short chain dehydrogenase/reductase family 9C, member 6
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: HSD17B6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat