Anti-MTHFD2 Rabbit Polyclonal Antibody
Catalog # 77743-587
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MTHFD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P13995
- Antigen synonyms:Bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase| mitochondrial| Descriptions| Methenyltetrahydrofolate cyclohydrolase| methylene tetrahydrofolate dehydrogenase (NAD+ dependent)| methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2| methylenetetrahydrofolate dehydrogenase (NADP+ dependent)| methylenetetrahydrofolate dehydrogenase 2| MGC82516| MTDC_HUMAN| mthfd2 methylene tetrahydrofolate dehydrogenase (NAD+ dependent)| NAD-dependent methylene tetrahydrofolate dehydrogenase| NAD-dependent methylenetetrahydrofolate dehydrogenase| NMDMC
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:Shipped on Blue ice
- Immunogen:NMDMC
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MTHFD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat