Anti-F8 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: F8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-070
- 77736-376
- Antigen symbol:
- F8
- F8
- Antigen synonyms:
- AHF| Antihemophilic factor| Coagulation factor VIII| coagulation factor VIII| procoagulant component| coagulation factor VIIIc| DXS1253E| F8| F8b| F8c| FA8_HUMAN| factor VIII F8B| Factor VIIIa light chain| FactorVIII| FVIII| Hema| Hemophilia A| Hemophilia| classic| OTTHUMP00000061446| OTTHUMP00000196174| Procoagulant component
- AHF|Antihemophilic factor|Coagulation factor VIII|coagulation factor VIII|procoagulant component|coagulation factor VIIIc|DXS1253E|F8|F8b|F8c|FA8_HUMAN|factor VIII F8B|Factor VIIIa light chain|FactorVIII|FVIII|Hema|Hemophilia A|Hemophilia|classic|OTTHUMP00000061446|OTTHUMP00000196174|Procoagulant component
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P00451
- P00451
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Coagulation factor VIII protein (1-216AA)
- Recombinant Human Coagulation factor VIII protein (1-216AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together