Anti-CD99 Rabbit Polyclonal Antibody
77739-518
New Product
- Antigen symbol:CD99
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P14209
- Antigen synonyms:Antigen identified by monoclonal 12E7| Y homolog| Antigen identified by monoclonal antibodies 12E7| F21 and O13| CD99 antigen| CD99 molecule| CD99_HUMAN| Cell surface antigen 12E7| Cell surface antigen HBA 71| Cell surface antigen O13| E2 antigen| HBA71| MIC 2X| MIC 2Y| MIC2 (monoclonal 12E7)| MIC2| MIC2X| MIC2Y| MSK5X| Protein MIC2| Surface antigen MIC2| T cell surface glycoprotein E2| T-cell surface glycoprotein E2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:Shipped on Blue ice
- Immunogen:Antigen identified by monoclonal 12E7
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IF: 1:200 to 1:1000, ELISA: 1:5000
Type:
Antigen: CD99
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human