Anti-MYH11 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: MYH11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-723
- 77860-189
- Antigen symbol:
- MYH11
- MYH11
- Antigen synonyms:
- AAT4|DKFZp686D10126|DKFZp686D19237|FAA4|FLJ35232|MGC126726|MGC32963|MYH 11|MYH11_HUMAN|Myosin 11|Myosin heavy chain 11|Myosin heavy chain 11 smooth muscle|Myosin heavy chain|Myosin heavy chain smooth muscle isoform|Myosin heavy polypeptide 11 smooth muscle|Myosin-11|SMHC|SMMHC|smooth muscle isoform|Smooth muscle myosin heavy chain 11 isoform SM2|Smooth muscle myosin heavy chain isoform SM2
- AAT4|DKFZp686D10126|DKFZp686D19237|FAA4|FLJ35232|MGC126726|MGC32963|MYH 11|MYH11_HUMAN|Myosin 11|Myosin heavy chain 11|Myosin heavy chain 11 smooth muscle|Myosin heavy chain|Myosin heavy chain smooth muscle isoform|Myosin heavy polypeptide 11 smooth muscle|Myosin-11|SMHC|SMMHC|smooth muscle isoform|Smooth muscle myosin heavy chain 11 isoform SM2|Smooth muscle myosin heavy chain isoform SM2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P35749
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Myosin-11 protein (1655-1797AA)
- Recombinant Human Myosin-11 protein (1655-1797AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together