Anti-GNRH2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: GNRH2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-252
- 77738-638
- Antigen symbol:
- GNRH2
- Antigen synonyms:
- GnRH II|GnRH-associated peptide 2|GnRH-associated peptide II|gnrh2|GON2_HUMAN|Gonadoliberin II|Gonadotropin-releasing hormone 2|Gonadotropin-releasing hormone II|LH-RH II|Luliberin II|Luteinizing hormone-releasing hormone II|Progonadoliberin 2|Progonadoliberin II
- GnRH II|GnRH-associated peptide 2|GnRH-associated peptide II|gnrh2|GON2_HUMAN|Gonadoliberin II|Gonadotropin-releasing hormone 2|Gonadotropin-releasing hormone II|LH-RH II|Luliberin II|Luteinizing hormone-releasing hormone II|Progonadoliberin 2|Progonadoliberin II
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O43555
- O43555
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Progonadoliberin-2 protein (24-120AA)
- Recombinant Human Progonadoliberin-2 protein (24-120AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together