Anti-FOSB Rabbit Polyclonal Antibody
Catalog # 77736-813
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:FOSB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P53539
- Antigen synonyms:Activator protein 1|AP 1|DKFZp686C0818|FBJ murine osteosarcoma viral oncogene homolog B|FOSB_HUMAN|G0/G1 switch regulatory protein 3|G0S3|GOS 3|GOS B|GOS3|GOSB|MGC42291|Oncogene FOS B|Oncogene Protein fos B|Protein fosB
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Activator protein 1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:2000, IHC:1:100 to 1:300, IF:1:200 to 1:1000, ELISA:1:5000
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human