Anti-NEK2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NEK2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-300
- 77744-301
- Antigen symbol:
- NEK2
- NEK2
- Antigen synonyms:
- HSPK 21|HsPK21|NEK 2|NEK2_HUMAN|NEK2A|Never in mitosis A-related kinase 2|NIMA (never in mitosis gene a) related kinase 2|NimA like protein kinase 1|NIMA related kinase 2|NimA related protein kinase 2|NimA-like protein kinase 1|NimA-related protein kinase 2|NLK 1|NLK1|Serine/threonine-protein kinase Nek2
- HSPK 21|HsPK21|NEK 2|NEK2_HUMAN|NEK2A|Never in mitosis A-related kinase 2|NIMA (never in mitosis gene a) related kinase 2|NimA like protein kinase 1|NIMA related kinase 2|NimA related protein kinase 2|NimA-like protein kinase 1|NimA-related protein kinase 2|NLK 1|NLK1|Serine/threonine-protein kinase Nek2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P51955
- P51955
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Serine/threonine-protein kinase Nek2 protein (1-135AA)
- Serine/threonine-protein kinase Nek2 protein (1-135AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together