Anti-HOXA9 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: HOXA9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-292
- 77857-575
- Antigen symbol:
- HOXA9
- HOXA9
- Antigen synonyms:
- ABD-B|Abd-B|drosophila|homolog of|D6a9|Homeobox 1G|Homeobox A9|Homeobox protein Hox-1.7|Homeobox protein Hox-1G|Homeobox protein Hox-A9|Homeodomain protein HOXA9|Hox-1.7|mouse|HOX1|Hox1.7|Hox1G|Hox1r5|Hoxa-9|Hoxa7|HOXA9|HOXA9/MSI2 fusion gene|included|HOXA9/NUP98 fusion gene|HXA9_HUMAN|MGC1934
- Hox1G|Hoxa7|Homeobox protein Hox-A9|HOXA9/MSI2 fusion gene|Abd-B|MGC1934|Homeobox A9|ABD-B|mouse|homolog of|HOXA9|Homeobox 1G|Homeobox protein Hox-1G|HOX1|Hoxa-9|Homeobox protein Hox-1.7|Homeodomain protein HOXA9|HOXA9/NUP98 fusion gene|D6a9|included|Hox1r5|HXA9_HUMAN|drosophila|Hox-1.7|Hox1.7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31269
- P31269
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Homeobox protein Hox-A9 protein (1-190AA)
- Recombinant Human Homeobox protein Hox-A9 protein (1-190AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together