Anti-GNL1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: GNL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77738-620
- 77856-243
- Antigen symbol:
- GNL1
- GNL1
- Antigen synonyms:
- GNL1_HUMAN|GTP binding protein HSR1|GTP-binding protein HSR1|Guanine nucleotide binding protein like 1|Guanine nucleotide-binding protein-like 1|HSR1|HSR1 GTP binding protein
- GNL1_HUMAN|GTP binding protein HSR1|GTP-binding protein HSR1|Guanine nucleotide binding protein like 1|Guanine nucleotide-binding protein-like 1|HSR1|HSR1 GTP binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P36915
- P36915
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Guanine nucleotide-binding protein-like 1 protein (251-500AA)
- Recombinant Human Guanine nucleotide-binding protein-like 1 protein (251-500AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together