Anti-ERAL1 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: ERAL1
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77854-862
- 77736-069
- Antigen symbol:
- ERAL1
- ERAL1
- Antigen synonyms:
- CEGA|Conserved ERA like GTPase|Conserved ERA-like GTPase|ERA |Era (E. coli G protein homolog) like 1|Era G protein like 1 (E. coli) |Era G protein like 1|ERA W|ERA-like protein 1|ERA-W|ERAL1_HUMAN|ERAL1A|ERAW|GTP binding protein era homolog|GTPase Era|mitochondrial|GTPase human homolog of E. coli essential cell cycle protein Era |H-ERA|HERA A|hERA|HERA B|HERAA|HERAB
- CEGA| Conserved ERA like GTPase| Conserved ERA-like GTPase| ERA | Era (E. coli G protein homolog) like 1| Era G protein like 1 (E. coli) | Era G protein like 1| ERA W| ERA-like protein 1| ERA-W| ERAL1_HUMAN| ERAL1A| ERAW| GTP binding protein era homolog| GTPase Era| mitochondrial| GTPase human homolog of E. coli essential cell cycle protein Era | H-ERA| HERA A| hERA| HERA B| HERAA| HERAB
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75616
- O75616
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human GTPase Era, mitochondrial protein (271-382AA)
- Recombinant Human GTPase Era, mitochondrial protein (271-382AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together