Anti-LPCAT2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: LPCAT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77859-104
- 77742-317
- Antigen symbol:
- LPCAT2
- LPCAT2
- Antigen synonyms:
- 1 acylglycerophosphocholine O acyltransferase|1 alkylglycerophosphocholine O acetyltransferase|1-acylglycerophosphocholine O-acyltransferase|1-alkylglycerophosphocholine O-acetyltransferase|A330042H22|Acetyl CoA:lyso PAF acetyltransferase|Acetyl-CoA:lyso-PAF acetyltransferase|Acetyl-CoA:lyso-platelet-activating factor acetyltransferase|acyl CoA:lysophosphatidylcholine acyltransferase 2|Acyl-CoA:lysophosphatidylcholine acyltransferase member 1|Acyltransferase like 1|Acyltransferase like 1A|Acyltransferase-like 1|AYTL1|Aytl1a|DKFZp686H22112|FLJ20481|lpafat1|LPC acyltransferase 2|LPCAT-2|Lyso PAF acetyltransferase|Lyso platelet activating factor (PAF) acetyltransferase|Lyso-PAF acetyltransferase|LysoPAFAT|LysoPAFAT/LysoPC acyltransferase 2|Lysophosphatidylcholine acyltransferase 2|OTTMUSP00000029641|PCAT2_HUMAN|RGD1563994
- 1 acylglycerophosphocholine O acyltransferase|1 alkylglycerophosphocholine O acetyltransferase|1-acylglycerophosphocholine O-acyltransferase|1-alkylglycerophosphocholine O-acetyltransferase|A330042H22|Acetyl CoA:lyso PAF acetyltransferase|Acetyl-CoA:lyso-PAF acetyltransferase|Acetyl-CoA:lyso-platelet-activating factor acetyltransferase|acyl CoA:lysophosphatidylcholine acyltransferase 2|Acyl-CoA:lysophosphatidylcholine acyltransferase member 1|Acyltransferase like 1|Acyltransferase like 1A|Acyltransferase-like 1|AYTL1|Aytl1a|DKFZp686H22112|FLJ20481|lpafat1|LPC acyltransferase 2|LPCAT-2|Lyso PAF acetyltransferase|Lyso platelet activating factor (PAF) acetyltransferase|Lyso-PAF acetyltransferase|LysoPAFAT|LysoPAFAT/LysoPC acyltransferase 2|Lysophosphatidylcholine acyltransferase 2|OTTMUSP00000029641|PCAT2_HUMAN|RGD1563994
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7L5N7
- Q7L5N7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lysophosphatidylcholine acyltransferase 2 protein (442-544AA)
- Recombinant Human Lysophosphatidylcholine acyltransferase 2 protein (442-544AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together