Anti-KDM5A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KDM5A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-546
- 77858-480
- Antigen symbol:
- KDM5A
- KDM5A
- Antigen synonyms:
- Histone demethylase JARID1A|Retinoblastoma binding protein 2|Jumonji/ARID domain-containing protein 1A|RBP2|Retinoblastoma-binding protein 2|RBBP2|Lysine-specific demethylase 5A|JARID1A|KDM5A_HUMAN|Jumonji/ARID domain containing protein 1A|RBBP-2
- Histone demethylase JARID1A|JARID1A|Jumonji/ARID domain containing protein 1A|Jumonji/ARID domain-containing protein 1A|KDM5A_HUMAN|Lysine-specific demethylase 5A|RBBP-2|RBBP2|RBP2|Retinoblastoma binding protein 2|Retinoblastoma-binding protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P29375
- P29375
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lysine-specific demethylase 5A protein (437-603AA)
- Recombinant Human Lysine-specific demethylase 5A protein (437-603AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together