Anti-GNA12 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GNA12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-157
- 77738-517
- Antigen symbol:
- GNA12
- GNA12
- Antigen synonyms:
- G alpha 12|G alpha-12|G-protein subunit alpha-12|gep|GNA12_HUMAN|guanine nucleotide binding protein (G protein) alpha 12|Guanine nucleotide binding protein alpha 12 subunit|Guanine nucleotide-binding protein subunit alpha-12|MGC104623|MGC99644|NNX3|RMP|WUGSC:H_GS165O14.2
- G alpha 12|G alpha-12|G-protein subunit alpha-12|gep|GNA12_HUMAN|guanine nucleotide binding protein (G protein) alpha 12|Guanine nucleotide binding protein alpha 12 subunit|Guanine nucleotide-binding protein subunit alpha-12|MGC104623|MGC99644|NNX3|RMP|WUGSC:H_GS165O14.2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q03113
- Q03113
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Guanine nucleotide-binding protein subunit alpha-12 protein (112-270AA)
- Recombinant Human Guanine nucleotide-binding protein subunit alpha-12 protein (112-270AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together