Anti-SLC25A1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: SLC25A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77799-880
- 77799-879
- Antigen symbol:
- SLC25A1
- SLC25A1
- Antigen name:
- Tricarboxylate transport protein, mitochondrial protein (144-182AA)
- Tricarboxylate transport protein, mitochondrial protein (144-182AA)
- Antigen synonyms:
- Citrate transport protein|CTP|mitochondrial|SLC20A3|solute carrier family 20 (mitochondrial citrate transporter), member 3|solute carrier family 25 (mitochondrial carrier, citrate transporter), member 1|Solute carrier family 25 member 1|Tricarboxylate carrier protein|Tricarboxylate transport protein|tricarboxylate transport protein, mitochondrial|TXTP_HUMAN
- Citrate transport protein|CTP|mitochondrial|SLC20A3|solute carrier family 20 (mitochondrial citrate transporter), member 3|solute carrier family 25 (mitochondrial carrier, citrate transporter), member 1|Solute carrier family 25 member 1|Tricarboxylate carrier protein|Tricarboxylate transport protein|tricarboxylate transport protein, mitochondrial|TXTP_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P53007
- P53007
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together