Anti-PDLIM7 Rabbit Polyclonal Antibody
77793-639
New Product
- Antigen name:Protein enigma
- Antigen symbol:PDLIM7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NR12
- Antigen synonyms:1110003B01Rik|Enigma|LIM domain protein|LIM domain protein enigma|LIM mineralization protein 1|Lim mineralization protein 3|LIM mineralization protein|LMP 1|LMP|LMP1|LMP3|PDLI7_HUMAN|PDLIM 7|Pdlim7|PDZ and LIM doamin protein 7|PDZ and LIM domain 7|PDZ and LIM domain protein 7|Protein enigma
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: PDLIM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat