Anti-RWDD2A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: RWDD2A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77841-604
- 77841-605
- Antigen symbol:
- RWDD2A
- RWDD2A
- Antigen name:
- RWD domain-containing protein 2A protein (1-292AA)
- RWD domain-containing protein 2A protein (1-292AA)
- Antigen synonyms:
- 1700030C20Rik|AI848608|dJ747H23.2|MGC13523|MGC138208|MGC144625|MGC144626|OTTHUMP00000016790|OTTHUMP00000040569|RWD domain containing 2|RWD domain containing 2A|RWD domain-containing protein 2A|RWD2A_HUMAN|Rwdd2|Rwdd2a|Rwdd2a RWD domain containing 2A
- 1700030C20Rik|AI848608|dJ747H23.2|MGC13523|MGC138208|MGC144625|MGC144626|OTTHUMP00000016790|OTTHUMP00000040569|RWD domain containing 2|RWD domain containing 2A|RWD domain-containing protein 2A|RWD2A_HUMAN|Rwdd2|Rwdd2a|Rwdd2a RWD domain containing 2A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UIY3
- Q9UIY3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together