Anti-PLXND1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PLXND1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-114
- 77826-115
- Antigen symbol:
- PLXND1
- PLXND1
- Antigen name:
- Plexin-D1 protein (1-222AA)
- Plexin-D1 protein (1-222AA)
- Antigen synonyms:
- KIAA0620|MGC75353|PLEX D1|PLEXD 1|PLEXD1|Plexin-D1|PlexinD1|PLXD1_HUMAN|PLXN D1|PLXND 1|PLXND1|PLXND1 protein
- KIAA0620|MGC75353|PLEX D1|PLEXD 1|PLEXD1|Plexin-D1|PlexinD1|PLXD1_HUMAN|PLXN D1|PLXND 1|PLXND1|PLXND1 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y4D7
- Q9Y4D7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together