Anti-POLR2D Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: POLR2D
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-741
- 77794-740
- Antigen symbol:
- POLR2D
- POLR2D
- Antigen name:
- DNA-directed RNA polymerase II subunit RPB4 protein (1-142AA)
- DNA-directed RNA polymerase II subunit RPB4 protein (1-142AA)
- Antigen synonyms:
- DNA directed RNA polymerase II 16 kDa polypeptide| DNA directed RNA polymerase II subunit D| DNA directed RNA polymerase II subunit RPB4| DNA-directed RNA polymerase II 16 kDa polypeptide| DNA-directed RNA polymerase II subunit D| DNA-directed RNA polymerase II subunit rpb4| HSRBP4| HSRPB4| polr2d| polymerase RNA II DNA directed polypeptide D| RBP4| RNA polymerase II 16 kDa subunit| RNA polymerase II subunit B4| RNA polymerase II subunit D| RNA polymerase II subunit hsRBP4| RPB16| RPB4_HUMAN
- DNA directed RNA polymerase II 16 kDa polypeptide| DNA directed RNA polymerase II subunit D| DNA directed RNA polymerase II subunit RPB4| DNA-directed RNA polymerase II 16 kDa polypeptide| DNA-directed RNA polymerase II subunit D| DNA-directed RNA polymerase II subunit rpb4| HSRBP4| HSRPB4| polr2d| polymerase RNA II DNA directed polypeptide D| RBP4| RNA polymerase II 16 kDa subunit| RNA polymerase II subunit B4| RNA polymerase II subunit D| RNA polymerase II subunit hsRBP4| RPB16| RPB4_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15514
- O15514
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together