Anti-SLCO1B3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLCO1B3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-870
- 77840-871
- Antigen symbol:
- SLCO1B3
- SLCO1B3
- Antigen name:
- Solute carrier organic anion transporter family member 1B3 protein (648-695AA)
- Solute carrier organic anion transporter family member 1B3 protein (648-695AA)
- Antigen synonyms:
- HBLRR|Liver-specific organic anion transporter 2|LST-2|LST-3TM13|LST3|OATP-8|OATP1B3|Organic anion transporter 8|Organic anion-transporting polypeptide 8|SLC21A8|SLCO1B3|SO1B3_HUMAN|Solute carrier family 21 member 8|Solute carrier organic anion transporter family member 1B3
- HBLRR|Liver-specific organic anion transporter 2|LST-2|LST-3TM13|LST3|OATP-8|OATP1B3|Organic anion transporter 8|Organic anion-transporting polypeptide 8|SLC21A8|SLCO1B3|SO1B3_HUMAN|Solute carrier family 21 member 8|Solute carrier organic anion transporter family member 1B3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NPD5
- Q9NPD5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together